Adapter, Ace-Thred, Column, Small Sample Injection, Ace Glass Incorporated
Supplier: Ace Glass
5826-07 5826-09 5826-11
89052-446EA 130.14 CAD
89052-446 89052-448 89052-450
Adapter, Ace-Thred, Column, Small Sample Injection, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This is a miniature column adapter for small sample injection.
  • Small sample injection
  • Threaded
  • Fabricated from borosilicate glass
Miniature column adapter is available with #7-#7 or #11-#11 Ace-Threds.