Column, Large Size, Epoxy Coated, wtih Ace-Threds, Ace Glass Incorporated
Supplier: Ace Glass
5820-105 5820-107 5820-109 5820-116 5820-119 5820-121 5820-125 5820-129 5820-133
89052-342EA 1784.22 CAD
89052-342 89052-344 89052-346 89052-348 89052-350 89052-354 89052-356 89052-358 89052-360
Column, Large Size, Epoxy Coated, wtih Ace-Threds, Ace Glass Incorporated
Chromatography Columns
This borosilicate glass chromatography column is used for liquid chromatography, ion-exchange, or activated carbon work.
The Ace-Thred ends are compatible with Ace-Thred fittings of the same size. These fittings can be used with easy-to-change support screens with a variety of openings never before offered.
Ordering information: For support stand, order 89076-384. Columns of larger int.Ø. and longer lengths can be fabricated, and most sizes can be jacketed as well. Contact your Avantor representative for further information.