Adapter, Michel-Miller, Injection Port, Ace Glass Incorporated
Supplier: Ace Glass
5807-14 5807-18 5807-48 5807-49
89052-138EA 306.15 CAD
89052-138 89052-140 89052-142 89052-144
Adapter, Michel-Miller, Injection Port, Ace Glass Incorporated
Fittings and Connectors
A simple, all-PTFE septum injector that allows for the most efficient use of Michel-Miller non-packed columns.
  • Direct on-column injection
  • Leak-tight seal without o-rings
  • Threaded
Injector allows direct on-column injection to the center of the filter disc at top of column without stopping the solvent flow. Designed to make a leak-tight seal without the use of o-rings. Simply tighten until white ring appears indicating a seal has been made, then secure locknut; no need to over tighten. Top thread is 5/16-inch-24 UNF for use with injection port cap that holds a septum. Male thread is size designation and matches thread size of columns. Bore is 1.5mm (.060-inches).
Ordering information: Adapter is available with or without Heli-Coil insert. Supplied with one PTFE coated silicone rubber septum. Use paper filter discs (not included) to retain packing material. Not suitable for larger chromatography columns. Contact your Avantor representative for details.