Axygen® Research-Grade Bevelled Reference Pipette Tips 200 µl, Corning
Supplier: Corning
TR-222-Y-R TR-222-Y-R-S TR-222-C-STK TR-222-Y TR-222-Y-STK TR-222-C-L-STK-S TR-222-C-STK-S
10011-376CS 1076.31 CAD
10011-376 10011-378 10011-894 10011-934 47734-722 47734-742 95042-896
Axygen® Research-Grade Bevelled Reference Pipette Tips 200 µl, Corning
Pipette Tips
Axygen tips are designed to fit a wide variety of single-channel and multichannel pipettors.
Flexible walls and a series of internal sealing rings ensure a secure fit and less force required to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry.