Axygen® Pipette Tip 200 µl with Graduation Marks, Corning
Supplier: Corning
TE-204-Y TE-204-Y-R TE-204-Y-L-R-S
80086-096CS 345.44 CAD
80086-096 80086-098 80086-106
Axygen® Pipette Tip 200 µl with Graduation Marks, Corning
Pipette Tips
Axygen brand 200 µl tips are designed to fit Eppendorf-Style single-channel and multi-channel pipettors.
Flexible walls and a series of internal sealing rings ensure a secure fit and less force required to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry. These tips have graduation marks at 10, 50 and 100 µl for your accurate pipetting.