Axygen® Pipette Tip 1200 µl with Graduation Marks, Corning
Supplier: Corning
TE-1004-B TE-1004-B-L-R TE-1004-B-L-R-S
80086-108CS 2362.05 CAD
80086-108 80086-116 80086-118
Axygen® Pipette Tip 1200 µl with Graduation Marks, Corning
Pipette Tips
Axygen brand tips are designed to fit Eppendorf-Style 1000 µl single-channel pipettors.
Flexible walls and a series of internal sealing rings ensure a secure fit and less force required to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry. These 1000 µl tip have graduation marks at 110 and 500 µl.