Axygen® Research-Grade Pipette Tips, Corning
Supplier: Corning
T-1000-C T-1000-C-R T-1000-C-R-S T-1005-WB-C
10011-306CS 661.12 CAD
10011-306 10011-308 10011-310 10011-916
Axygen® Research-Grade Pipette Tips, Corning
Pipette Tips
These tips are designed to fit a wide variety of 1000 µl single-channel pipettors.
Flexible walls and a series of internal sealing rings ensure a secure fit and less force required to load and eject. Accuracy and precision testing is performed for every manufacturing lot using the finest pipettors in the industry. Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures.