HyClone™ Amino Acids, HyClone products (Cytiva)
Supplier: Cytiva
SH30034.02
CA16777-162EA 40.12 CAD
CA16777-162 CA82007-322
HyClone™ Amino Acids, HyClone products (Cytiva)
Cell Culture Supplements and Additives
HyClone Amino Acids are ideal for supplementation of serum- and protein-free media
They support the growth of cells that have high energy demands and synthesize large amounts of proteins and nucleic acids.