Barrier Pipette Tips, Sorenson BioScience
Supplier: Sorenson Bioscience
30550 30550T 30510-X 30510T-X 34000
89082-364CS 905.22 CAD
89082-364 89082-366 89082-368 89082-370 89082-372
Barrier Pipette Tips, Sorenson BioScience
Pipette Tips
The barrier pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment and feature an inert hydrophobic aerosol-blocking barrier.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips minimize sample retention.
Order Now
SPECIFICATIONS
Environmentally Preferable