Pipette Tips, Sorenson BioScience
Supplier: Sorenson BioScience
30140 30800T 30920T 30430 27120 30800 30170 23570 30460 34750 23580 30160 30920 30930 34770
15001-099CS 506.43 CAD
15001-099 89082-382 89082-376 89082-386 89082-374 14214-395 53550-073 89082-380 53550-067 89082-390 13503-464 53550-071 14006-902 14006-914 89082-378
Pipette Tips, Sorenson BioScience
Pipette Tips
Sorenson BioScience universal fit pipette tips are manufactured from medical grade polypropylene in a tightly controlled production environment.
Certified free of RNase / DNase, Human DNA, Pyrogens and PCR Inhibitors.
The accurate and precise tips are designed to fit most research-grade pipettors. Available in low binding, pipette tips reduce sample retention.