Axygen® Screw Top Microcentrifuge Tubes with Caps, Corning
Supplier: Corning
SCT-200-SSRS SCT-200-SSYS SCT-200-SSBS SCT-200-SS-O-S SCT-050-B-S SCT-050-G-S SCT-050-O-S SCT-050-R-S SCT-050-SS-BS SCT-050-SS-G-S SCT-050-SS-O-S SCT-050-SS-R-S SCT-050-SS-W-S SCT-050-SS-Y-S SCT-050-Y-S SCT-150-B-S SCT-150-O-S SCT-150-R-S SCT-150-SS-B-S SCT-150-SS-G-S SCT-150-SS-R-S SCT-150-Y-S SCT-200-SS-V-S SCT-45-C-S SCT-45-SS-C-S
47742-966CS 4251.19 CAD
47742-966 47743-808 47743-810 89139-866 89179-682 89179-686 89179-688 89179-692 89179-696 89179-700 89179-702 89179-704 89179-708 89179-710 89179-718 89179-722 89179-726 89179-728 89179-730 89179-734 89179-738 89179-752 89179-766 89179-774 89179-778
Axygen® Screw Top Microcentrifuge Tubes with Caps, Corning
Tubes Microcentrifuge Tubes
These conical-bottom screw cap microcentrifuge tubes serve a broad range of laboratory needs including collection, pressing, centrifugation, long-term storage, and OEM packaging and shipping of restriction enzymes or other DNA/RNA modifying reagents.
Microcentrifuge tubes serve a broad range of laboratory needs including collection, centrifugation, long-term storage, and OEM packaging and shipping of restriction enzymes or other DNA/RNA modifying reagents. Ribs on cap and tube are designed to fit the AxyRack™ microtube racks (see 10011-272 series) to allow single-handed opening and closing. MAXYMum Recovery™ tubes feature an ultra-smooth surface completely free of occlusions and cavities, virtually eliminating sample retention while improving accuracy. Certified RNase-, DNase-, and pyrogen-free.
Color-coded caps provide a syringe port that is easily pierceable for radioisotope labeling and fraction collecting. The threaded O-ring design ensures a uniform closure allowing centrifugation up to 20000×g. Microcentrifuge tubes have ribs on the cap and tube to fit the AxyRack™ microtube racks (see 10011-272 series) to allow single-handed opening and closing. Tubes without ribs are also available; contact your sales representative for information.