Axygen® Robotic Pipette Tips, PK Series, Corning
Supplier: Corning
PK-384-R PK-384-R-S PKF-384-R-S
80086-038CS 6302.03 CAD
80086-038 80086-040 80086-042
Axygen® Robotic Pipette Tips, PK Series, Corning
Pipette Tips
The PK series robotic tips for the PerkinElmer® and Packard® Systems are certified RNase-, DNase-, and pyrogen-free.
The PK series robotic tips are engineered to exact dimensions of OEM tips to ensure automation compatibility.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Reynosa