HyClone™ Newborn Bovine Calf Serum, US Origin, Cytiva
Supplier: Cytiva
3011803IR25-40 SH30118.02 SH30118.02HI SH30118.03 SH30118.03HI SH30118.04 SH30118.04HI
CA76324-894EA 202.99 CAD
CA76324-894 CA16777-220 CA76236-316 CA16777-222 CA76236-318 CA82007-448 CA76236-322
HyClone™ Newborn Bovine Calf Serum, US Origin, Cytiva
Sera (Serum)
Bovine calf serum products are excellent and cost-effective alternatives to fetal bovine serum (FBS). Bovine calf serum products contain exceptionally high levels of transferrin, which can provide three to four times as much available iron as FBS.
  • Complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
HyClone™ Newborn Bovine Calf Serum, US Origin undergoes the same careful collection and processing procedures (including venipuncture) used for our bovine serum products. Newborn calf serum is sourced from animals that are typically less than 10 days old and filtered through three sequential 100 nm (0.1 μm) pore sized-rated filters.