Automation Pipette Tips for Caliper Workstations, Thermo Scientific
Supplier: Thermo Scientific
160-R-960S 920-261 920-262 920-021
14004-636CS 3014.71 CAD
14004-636 72830-600 72830-602 72830-604
Automation Pipette Tips for Caliper Workstations, Thermo Scientific
Pipette Tips
Thermo scientific automation pipette tips made to fit the Caliper RapidPlate, Presto, Sciclone, Zephry, and Allegro Automated Workstations.
It provides a solution for your Beckman robotic pipette tips. These automation pipette tips have been designed, validated, and tested on a Caliper robot to ensure equal performance without the need to modify your existing protocols and programs. Our low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Each lot must pass a 15-point QC inspection including performance testing on our Caliper workstation before tips are released. All tips are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.