Water, HyClone™ HyPure, Molecular Biology Grade

Supplier: Cytiva

SH30538.FS SH30538.LS SH30538.01 SH30538.02 SH30538.03
CA16750-104PK 156.33 CAD
CA16750-104 CA16750-128 CA82007-332 CA82007-334 CA82007-336
Water, HyClone™ HyPure, Molecular Biology Grade
Water

Nuclease-free HyClone Molecular Biology-Grade Water, containing no DEPC or other carcinogenic chemicals is used for critical applications involving nucleotides such as RNA and DNA.


  • Significantly low levels of endotoxins
  • Certified to have no detectable activity in DNase, RNase, and protease assays
  • Maximum allowable limit of endotoxin is 0.05 EU/mL


HyClone HyPure Molecular Biology Grade Water purification process includes continuous deionization, reverse osmosis, ultraviolet light treatment, distillation, and 0.1 µm filtration.


Certifications: Manufactured using ISO 9001 and ISO 13485 certified processes according to 21 CFR 820 medical device current good manufacturing practices (cGMP).filtration

Formula: H₂O
Melting Pt: 0 °C
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Specification Test Results

Endotoxin Concentration ≤ 0.05 EU/mL
Sterilisation 0.1 µm sterile filtered
pH 5.0 to 7.0
Storage 2° to 30° C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR