HyClone™ Phosphate Buffered Saline (PBS), HyClone products (Cytiva)
Supplier: Cytiva
SH30256.LS SH30258.01 SH30256.01 SH30256.02
CA16750-078PK 172.38 CAD
CA16750-078 CA16750-122 CA16777-250 CA16777-252 CA16777-251 CA16777-249
HyClone™ Phosphate Buffered Saline (PBS), HyClone products (Cytiva)
Buffers Cell Culture Buffers
Balanced salt solution used for multiple cell culture applications.
  • Full traceability and documented origin of all formula ingredients
  • Validated and consistent production processes
0.1 µm sterile filtered
Endotoxin Concentration: ≤1.0 EU/mL
Certifications: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities