HyClone™ Dulbecco's Phosphate Buffered Saline Solution, HyClone products (Cytiva)

Supplier: Cytiva

SH30264.01 SH30264.02 SH30264.FS SH30028.01 SH30028.02 SH30378.02 SH30028.03 SH30028.04 SH30028.FS
CA16777-257EA 30.48 CAD
CA16777-257 CA16777-259 CA16750-102 CA82013-222 CA82020-066 CA16777-644 CA82013-318 CA16777-150 CA95059-620 CA82024-294 CA16750-076 CA16750-110
HyClone™ Dulbecco's Phosphate Buffered Saline Solution, HyClone products (Cytiva)
Buffers Cell Culture Buffers

Balanced salt solution used for multiple cell culture applications, (liquid form).


  • Certificate of Analysis included with product and available electronically for future reference.
  • Standard stock items from 100 mL to 200 L
  • Volumes up to 1000 L available as made to order
  • Available in 1X and 10X concentrations to meet specific needs


For making isotonic solutions for cell or tissue rinsing, cell dilution, reagent prep, transportation etc. where maintanence of cell integrity is required for further manufacturing or research use.


Certifications: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.


Caution: Not for diagnostic or therapeutic use.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR