Flask, Shaker, Four Bottom Baffles, Ace Glass Incorporated
Supplier: Ace Glass
3893-16 3893-07 3893-12
71000-402CS 950.11 CAD
71000-402 71000-436 71000-470
Flask, Shaker, Four Bottom Baffles, Ace Glass Incorporated
Flasks Cell Culture Flasks
This flask shaker flask has with four bottom baffles
Long-style, 38 mm O.D. neck. Used for converting homogenous tissue samples into cell suspensions by digestion of connective tissues. The constricted neck helps reduce spillage and foaming. Fabricated from borosilicate glass.
Ordering information: Closures not included.
Order Now
SPECIFICATIONS
Environmentally Preferable