Flask, Shaker, Beaded Neck, Ace Glass Incorporated
Supplier: Ace Glass
3889-24 3889-14 3889-29 3889-18 3889-22
71000-390CS 863.93 CAD
71000-390 71000-404 71000-412 71000-424 71000-458
Flask, Shaker, Beaded Neck, Ace Glass Incorporated
Flasks Cell Culture Flasks
Shake style culture flask with beaded neck and three bottom edge baffles to enhance mixing and oxygen transfer to media.