Flask, Culture, Screw Cap, Ace Glass Incorporated
Supplier: Ace Glass
3877-30 3877-45
89083-532EA 535.47 CAD
89083-532 89083-524
Flask, Culture, Screw Cap, Ace Glass Incorporated
Flasks Cell Culture Flasks
This wide mouth flask is designed for culturing organisms requiring a large surface-area-to-volume ratio
It has triple baffles on the bottom outside edges to achieve maximal oxygen transfer to culture medium. Fabricated from borosilicate glass.
Ordering information: Supplied with GL45 polypropylene cap. Caution! Do not place on direct heat source.