Stainless Steel Closures, Ace Glass Incorporated
Supplier: Ace Glass
3918-50 3918-53 3918-59 3918-51 3918-56 3918-52 3918-54 3918-61 3918-55 3918-58 3918-57
71000-332CS 1233.33 CAD
71000-332 71000-340 71000-330 71000-324 71000-336 10098-726 71000-322 71000-334 71000-328 71000-326 71000-338
Stainless Steel Closures, Ace Glass Incorporated
Closures Test/Sample Tube Closures
These closures are designed for use with culture tubes and flasks and are available with or without gripping finger indents
Closures are made of 305 Stainless Steel and are fully autoclavable.