Filling Bell Aseptic, Ace Glass Incorporated
Supplier: Ace Glass
3868-10 3868-02 3868-05
71000-312CS 592.07 CAD
71000-312 71000-314 71000-316 71000-318 71000-320
Filling Bell Aseptic, Ace Glass Incorporated
Bottles Cell Culture Bottles
This apparatus is used for aseptic filling of vessels.
  • Fabricated from borosilicate glass
Rubber tubing is attached to the top hose connection, the receiving vessel is placed inside the bell.