Flask, Shaker, Three Extra Deep Baffles, Ace Glass Incorporated
Supplier: Ace Glass
3887-07 3887-22 3887-24 3887-17 3887-05 3887-11 3887-13 3887-09 3887-19
71000-398CS 712.35 CAD
71000-398 71000-420 71000-450 71000-440 71000-410 71000-454 71000-476 71000-432 71000-466
Flask, Shaker, Three Extra Deep Baffles, Ace Glass Incorporated
Flasks Cell Culture Flasks
This shaker flask has a long-style neck with three extra deep baffles to enhance gas transfer when used with a rotary or reciprocating shakers
The long neck reduces splashing, and is designed for polypropylene closures. Fabricated from borosilicate glass.
Ordering information: Closure NOT included.