Flask, Shaker, Three Side Baffles, Ace Glass Incorporated
Supplier: Ace Glass
3883-13 3883-05 3883-15 3883-07 3883-09 3883-11 3883-03
71000-394CS 727.96 CAD
71000-394 71000-406 71000-416 71000-428 71000-446 71000-462 71000-472
Flask, Shaker, Three Side Baffles, Ace Glass Incorporated
Flasks Cell Culture Flasks
These long-style neck shaker flasks have three side baffles that enhance mixing and gas transfer when used with rotary or reciprocating shakers
Flasks can be used for cell culture applications. The long neck reduces splashing, and is designed for polypropylene or stainless steel closures. Fabricated from borosilicate glass.
Ordering information: Polypropylene or stainless steel closures are available separately.