Flask, Fernbach, Culture, Ace Glass Incorporated
Supplier: Ace Glass
3879-18 3879-30 3879-34
71000-346CS 1241.53 CAD
71000-346 71000-352 89215-032
Flask, Fernbach, Culture, Ace Glass Incorporated
Flasks Cell Culture Flasks
This Fernbach culture flask is designed for culturing organisms requiring a large surface-to-volume ratio
It has a long, 38 mm neck that takes stainless steel or plastic closures. It does not have baffles. Fabricated from borosilicate glass.
Ordering information: Closure NOT included.