Buret, Both Ends Open, Ace Glass Incorporated
Supplier: Ace Glass
5758-04 5758-08 5758-10
89051-928EA 228.33 CAD
89051-928 89051-930 89051-932
Buret, Both Ends Open, Ace Glass Incorporated
Burettes
This is a graduated, straight-through buret, open at top and bottom
Top is beaded, bottom is fire polished. Can be used in the fabrication of apparatus requiring graduation. Fabricated from borosilicate glass.