Tissue Homogenizer, Tenbroeck, Ace Glass Incorporated
Supplier: Ace Glass
8358-15 8358-05 8358-10
71000-534CS 620.36 CAD
71000-534 71000-538 71000-542
Tissue Homogenizer, Tenbroeck, Ace Glass Incorporated
Homogenizers Tissue Grinders
These all-glass Tenbroeck tissue grinders come with precision-made, interchangeable pestles and tubes
Cited in Biochemistry Laboratory, 2nd Edition, by Rodney Boyer, "Tools for the Preparation of Crude Cell Extracts".
The hollow handle permits packing with ice. This unit also features an expanded reservoir and a pouring lip. Designed for tissues such as liver, intestines, and heart. Fabricated from borosilicate glass.