Tissue Homogenizer, Potter-Elehjem, Ace Glass Incorporated
Supplier: Ace Glass
8355-10 8355-15 8355-02 8355-30 8355-55 8355-05
71000-522CS 692.29 CAD
71000-522 71000-524 71000-526 71000-528 71000-530 71000-532
Tissue Homogenizer, Potter-Elehjem, Ace Glass Incorporated
Homogenizers Tissue Grinders
When using this Potter-Elehjem tissue homogenizer, homogenization occurs as the sample and buffer are forced through the cylindrical portion of the mortar as the pestle is rotated downward
Cited in Biochemistry Laboratory, 2nd Edition, by Rodney Boyer, "Tools for the Preparation of Crude Cell Extracts".
The mortar is not ground, and the pestle contact surface is PTFE, threaded onto a stainless steel shaft. Tube fabricated from borosilicate glass.
Order Now
SPECIFICATIONS
Environmentally Preferable