Tissue Homogenizer, Dounce, Ace Glass Incorporated
Supplier: Ace Glass
8343-15 8343-01 8343-40 8343-07
71000-512CS 855.03 CAD
71000-512 71000-514 71000-516 71000-518
Tissue Homogenizer, Dounce, Ace Glass Incorporated
Homogenizers Tissue Grinders
This Dounce tissue homogenizer is supplied with two pestles, one "loose" and one "tight", to ensure dissociation of cells into fine particles with minimal damage to cell nuclei
Cited in Biochemistry Laboratory, 2nd Edition, by Rodney Boyer, "Tools for the Preparation of Crude Cell Extracts".
Use the large-clearance pestle for initial reduction of soft tissue. Complete the homogenization with the small-clearance pestle. Particularly useful in enzyme studies where heat build-up must be avoided. Fabricated from borosilicate glass.
Order Now
SPECIFICATIONS
Environmentally Preferable