Acetic acid 0.0100 N (N/100)

Supplier: Ricca Chemical

R0143000-500A R0143000-4A
RC143-16EA 75.84 CAD
RC143-16 RC143-1
Acetic acid 0.0100 N (N/100)
Acetic acid
Formula: H₃CCOOH
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, no odor
Assay at 20 °C (traceable to NIST SRM 84) 0.01000 ± 0.00005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR