Acetic acid 10% (v/v) in aqueous solution (1 + 9)

Supplier: Ricca Chemical
Danger

R0135000-4A R0135000-1A R0135000-500A R0135000-20F R0135000-55E
RC135-1EA 224.36 CAD
RC135-1 RC135-32 RC135-16 RCR0135000-20F RCR0135000-55E
Acetic acid 10% (v/v) in aqueous solution (1 + 9)
Acetic acid
Formula: H₃CCOOH
Melting Pt: ∼ 0°C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2790
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Thymol Blue) 9.94-10.06 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR