Silver nitrate 1.00 N in aqueous solution
Supplier: Ricca Chemical
|
Danger
|
R7025000-500B
R7025000-1B
RC702516EA
1459.92
CAD
RC702516
RC702532
Silver nitrate 1.00 N in aqueous solution
Silver nitrate
Formula:
AgNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 EINECS: 231-853-9 UN: 3264 ADR: 8,II |
|
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 999) | 1.000 ± 0.001 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...