Silver nitrate 0.500 N (N/2) in aqueous solution

Supplier: Ricca Chemical
Danger

R7020000-4B R7020000-500B R7020000-1B
RC70201EA 4261.26 CAD
RC70201 RC702016 RC702032
Silver nitrate 0.500 N (N/2) in aqueous solution
Silver nitrate
Formula: AgNO₃
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3264
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.5000 ± 0.0002 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR