Silver nitrate 0.0855 N in aqueous solution
Supplier: Ricca Chemical
|
Warning
|
R6980000-4B
R6980000-20F
RC69801EA
1531.02
CAD
RC69801
RC6980-5
Silver nitrate 0.0855 N in aqueous solution
Silver nitrate
Formula:
AgNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 EINECS: 231-853-9 UN: 3082 ADR: 9,III |
|
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 999) | 0.08550 ± 0.00005 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...