Silver nitrate 0.0500 N (N/20) in aqueous solution analytical reagent
Supplier: Ricca Chemical
Synonyms:
Lunar caustic, Silver mononitrate, Nitric acid silver salt
R6970000-4B
R6970000-1B
RC69701EA
722.06
CAD
RC69701
RC697032
Silver nitrate 0.0500 N (N/20) in aqueous solution analytical reagent
Silver nitrate
Formula:
AgNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 EINECS: 231-853-9 |
|
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 999) | 0.05000 ± 0.00005 N at 20 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...