Silver nitrate 0.0250 N (N/40) in aqueous solution analytical reagent

Supplier: Ricca Chemical

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

R6920000-120B R6920000-1B
RC6920-4EA 97.96 CAD
RC6920-4 RC6920-32
Silver nitrate 0.0250 N (N/40) in aqueous solution analytical reagent
Silver nitrate
Formula: AgNO₃
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.02500 ± 0.00001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR