Sulfuric acid 6.00 N in aqueous solution

Supplier: Ricca Chemical
Danger

R8330000-1A R8330000-500A R8330000-20F R8330000-4A R8330000-2.5D
RC833032EA 162.74 CAD
RC833032 RC833016 RC8330-5 RC8330-1 RC8330-5PT
Sulfuric acid 6.00 N in aqueous solution
Sulfuric acid
Formula: H₂SO₄
Density: 1.146 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 6.000 ± 0.005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR