Sulfuric acid 4.00 N in aqueous solution
Supplier: Ricca Chemical
|
Danger
|
R8322000-4A
R8322000-1A
R8322000-20F
R8322000-500A
RC8322-1EA
241.74
CAD
RC8322-1
RC8322-32
RC83225
RCR8322000500A
Sulfuric acid 4.00 N in aqueous solution
Sulfuric acid
Formula:
H₂SO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 2796 ADR: 8,II |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sodium Hydroxide/Phenolphthalein) | 3.999-4.001 N at 20°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...