Sodium chloride 25% (w/v) in aqueous solution
Supplier: Ricca Chemical
R7219500-4A
R7219500-1A
RC7219.5-1EA
372.88
CAD
RC7219.5-1
RC7219.5-32
Sodium chloride 25% (w/v) in aqueous solution
Sodium chloride
Formula:
NaCl Density: 1…1.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 999) | 25.00 ± 0.07 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...