Sulfuric acid 5.00 N in aqueous solution
Supplier: Ricca Chemical
|
Danger
|
R8325000-500A
R8325000-4A
R8325000-20F
R8325000-1A
RC832516EA
96.38
CAD
RC832516
RC8325-1
RC8325-5
RCR8325000-1A
Sulfuric acid 5.00 N in aqueous solution
Sulfuric acid
Formula:
H₂SO₄ Density: 1.146 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 2796 ADR: 8,II |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sodium Hydroxide/Phenolphthalein) | 4.995-5.005 N at 20°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...