Sulfuric acid 1.00 N in aqueous solution

Supplier: Ricca Chemical

R8300000-10F R8300000-1A R8300000-20F R8300000-4A
RC830025EA 301.78 CAD
RC830025 RC830032 RC83005 RC83001
Sulfuric acid 1.00 N in aqueous solution
Sulfuric acid
Formula: H₂SO₄
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 1.000 ± 0.001 N at 20.0 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR