Arsenic(III) oxide standard 0.100 N (N/10)

Supplier: Ricca Chemical

R0820000-500A R0820000-4A
RC82016EA 145.36 CAD
RC82016 RC820-1
Arsenic(III) oxide standard 0.100 N (N/10)
Arsenic (III) oxide

Arsenite Standard, RICCA Reagents for Published Test Methods


Contains:
Water ACS/ASTM/USP/EP
Arsenic Trioxide 1327-53-3 ACS
Sodium Hydroxide 1310-73-2 ACS
Sodium Bicarbonate 144-55-8

Formula: As₂O₃
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003433
CAS Number: 1327-53-3
EINECS: 231-639-5
UN: 1561
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR