Sodium thiosulfate 1.00 N in aqueous solution
Supplier: Ricca Chemical
Synonyms:
di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite
R7970000-4A
R7970000-500A
R7970000-1A
RC79701EA
347.6
CAD
RC79701
RC797016
RC797032
Sodium thiosulfate 1.00 N in aqueous solution
Sodium thiosulphate
Formula:
Na₂S₂O₃ MW: 158.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003499 CAS Number: 7772-98-7 EINECS: 231-867-5 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 136) | 1.000 ± 0.001 N at 20.0 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...