Sodium thiosulfate 0.500 N (N/2) aqueous solution analytical reagent

Supplier: Ricca Chemical

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

R7966000-4A R7966000-500A R7966000-1A
RC7966-1EA 316 CAD
RC7966-1 RC7966-16 RC7966-32
Sodium thiosulfate 0.500 N (N/2) aqueous solution analytical reagent
Sodium thiosulphate
Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.5000 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR