Sodium thiosulfate 0.200 N (N/5) in aqueous solution

Supplier: Ricca Chemical

R7960000-20F R7960000-4A R7960000-1A
RC7960-5EA 393.42 CAD
RC7960-5 RC7960-1 RC7960-32
Sodium thiosulfate 0.200 N (N/5) in aqueous solution
Sodium thiosulphate
Formula: Na₂S₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.2000 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR