Sodium thiosulfate 0.250 N (N/4) in aqueous solution

Supplier: Ricca Chemical

R7961000-4A R7961000-20F
RC7961-1EA 255.96 CAD
RC7961-1 RC7961-5
Sodium thiosulfate 0.250 N (N/4) in aqueous solution
Sodium thiosulphate
Formula: Na₂S₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.2500 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR