Sodium chloride 20% w/v aqueous solution analytical reagent

Supplier: Ricca Chemical

Synonyms: Saline, NaCl, Sea salt, Halite

R7218600-500A R7218600-1A
RC7218.6-16EA 101.12 CAD
RC7218.6-16 RC7218.6-32
Sodium chloride 20% w/v aqueous solution analytical reagent
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Density: 1…1.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay 20.0 ± 0.3 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR