Sodium thiosulfate 0.100 N (N/10) in aqueous solution
Supplier: Ricca Chemical
Synonyms:
di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite
R7950000-4A
R7950000-10F
R7950000-20F
R7950000-1A
RC79501EA
238.58
CAD
RC79501
RC795025
RC79505
RC795032
Sodium thiosulfate 0.100 N (N/10) in aqueous solution
Sodium thiosulphate
Formula:
Na₂S₂O₃ MW: 158.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003499 CAS Number: 7772-98-7 EINECS: 231-867-5 |
Specification Test Results
Appearance | Clear, colorless, slight organic odor |
Assay at 20 °C (traceable to NIST SRM 136) | 0.1000 ± 0.0001 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...