Sodium thiosulfate 0.0500 N (N/20) in aqueous solution

Supplier: RICCA CHEMICAL CO CA

Synonyms: di-Sodium thiosulfateSodium hyposulphitedi-Sodium thiosulphateSodiumthiosulfateSodium hyposulfiteAntichlorSodiumthiosulphate

R7940000-1A R7940000-20F R7940000-4A R7940000-1A
CARC7940-32EA 88.42 CAD
RC7940-32 CARC7940-32 RC-7940-1 RC7940-5
Sodium thiosulfate 0.0500 N (N/20) in aqueous solution
Sodium thiosulphate
Formula: Na₂S₂O₃
MW: 158.11 g/mol
MDL Number: MFCD00003499
CAS Number: 7772-98-7

Order Now

Specification Test Results

Appearance Clear, colorless, slight organic odor
Assay at 20 °C (traceable to NIST SRM 136) 0.05000 ± 0.00002 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR