Sodium hydroxide 5.00 N in aqueous solution
Supplier: Ricca Chemical
|
Danger
|
R7465000-4A
R7465000-1A
R7465000-10F
RC7465-1EA
303.36
CAD
RC7465-1
RC746532
RC7465-2.5
Sodium hydroxide 5.00 N in aqueous solution
Sodium hydroxide
Formula:
NaOH Boiling Pt: 109 °C (1013 hPa) Density: ≤1.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 723) | 5.000 ± 0.005 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...